missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADAL Protein
A cDNA sequence encoding the ADAL was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10960-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
161823 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEFMIEAE EQQPCKTDFY SELPKVELHA HLNGSISSHT MKKLIAQKPD LKIHDQMTVI DKGKKRTLEE CFQMFQTIHQ LTSSPEDILM VTKDVIKEFA DDGVKYLELR STPRRENATG MTKKTYVESI LEGIKQSKQE NLDIDVRYLI AVDRRGGPLV AKETVKLAEE FFLSTEGTVL GLDLSGDPTV GQAKDFLEPL LEAKKAGLKL ALHLSEIPNQ KKETQILLDL LPDRIGHGTF LNSGEGGSLD LVDFVRQHRI PLGKAWSFRS SR | |
Greater than 80% as determined by Analysis by SDS-PAGE. |
ADAL Protein | |
Research Use Only | |
ADAL | |
Human | |
His | |
The ADAL solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |