missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Activin B Protein
A cDNA sequence encoding the Activin B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10504-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Activin B Protein | |
Research Use Only | |
Recombinant Protein | |
Plants | |
HHHHHHHHHH GLECDGRTNL CCRQQFFIDF RLIGWNDWII APTGYYGNYC EGSCPAYLAG VPGSASSFHT AVVNQYRMRG LNPGTVNSCC IPTKLSTMSM LYFDDEYNIV KRDVPNMIVE ECG | |
Greater than 97.0% as determined by Analysis by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
His | |
Lyophilized from 1 mg/ml solution in 0.05M Tris-HCl buffer pH 7.4. |