missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACTA2 Protein
A cDNA sequence encoding the ACTA2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10949-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
ACTA2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMEEEDS TALVCDNGSG LCKAGFAGDD APRAVFPSIV GRPRHQGVMV GMGQKDSYVG DEAQSKRGIL TLKYPIEHGI ITNWDDMEKI WHHSFYNELR VAPEEHPTLL TEAPLNPKAN REKMTQIMFE TFNVPAMYVA IQAVLSLYAS GRTTGIVLDS GDGVTHNVPI YEGYALPHAI MRLDLAGRDL TDYLMKILTE RGYSFVTTAE REIVRDIKEK LCYVALDFEN EMATAASSSS LEKSYELPDG QVITIGNERF RCPETLFQPS FIGMESAGIH ETTYNSIMKC DIDIRKDLYA NNVLSGGTTM YPGIADRMQK EITALAPSTM KIKIIAPPER KYSVWIGGSI LASLSTFQQM WISKQEYDEA GPSIVHRKCF | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ACTA2 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |