missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L10a Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L10a, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8500-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_009035.3 | |
60S ribosomal protein L10a Protein | |
Research Use Only | |
RPL10A | |
Recombinant Protein | |
E. coli | |
KVSRDTLYEA VREVLHGNQR KRRKFLETVE LQISLKNYDP QKDKRFSGTV RLKSTPRPKF SVCVLGDQQH CDEAKAVDIP HMDIEALKKL NKNKKLVKKL AKKYDAFLAS ESLIKQIPRI LGPGLNKAGK FPSLLTHNEN MVAKVDEVKS TIKFQMKKVL CLAVAVGHVK MTDDELVYNI HLAVNFLVSL LKKNWQNVRA LYIKSTMGKP QR | |
Greater than 90% as determined by SDS-PAGE. |
4736 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |