missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 40S ribosomal protein S10 Protein
A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S10, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8417-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001005.1 | |
40S ribosomal protein S10 Protein | |
Research Use Only | |
RPS10 | |
Recombinant Protein | |
E. coli | |
PKKNRIAIYE LLFKEGVMVA KKDVHMPKHP ELADKNVPNL HVMKAMQSLK SRGYVKEQFA WRHFYWYLTN EGIQYLRDYL HLPPEIVPAT LRRSRPETGR PRPKGLEGER PARLTRGEAD RDTYRRSAVP PGADKKAEAG AGSATEFQFR GGFGRGRGQP PQ | |
Greater than 90% as determined by SDS-PAGE. |
6204 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |