missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV HTLV-1 gp46 mosaic Protein
A cDNA sequence encoding the HTLV-1 gp46 mosaic was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12355-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HIV HTLV-1 gp46 mosaic Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged |
HTLV-1 gp46 protein is >95% pure as determined by 10% SDS-PAGE (coomassie staining) and RP-HPLC. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM NaPO4 pH 6.0, 0.1% SDS, 1mM DTT, 1mM EDTA. |