missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBV HBe Protein
A cDNA sequence encoding the HBV HBe was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12132-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Hepatitis B HBV HBe Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/ml) 50mM Tris-HCl pH 8.5, 5mM DTT, 10mM BMe and 8M Urea. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
GST | |
HBV HBe protein is >95% pure as determined by 10% PAGE (coomassie staining). |