missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant H. pylori Vacuolating cytotoxin autotransporter Protein
A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Vacuolating cytotoxin autotransporter, was expressed in the hosts and tags indicated. option, abo
Marca: enQuireBio™ QP7226-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_207680.1 | |
Vacuolating cytotoxin autotransporter Protein | |
Research Use Only | |
vacA | |
Recombinant Protein | |
E. coli | |
KYEKPTNVWA NAIGGTSLNN GGNASLYGTS AGVDAYLNGE VEAIVGGFGS YGYSSFNNQA NSLNSGANNT NFGVYSRIFA NQHEFDFEAQ GALGSDQSSL NFKSALLRDL NQSYNYLAYS AATRASYGYD FAFFRNALVL KPSVGVSYNH LGSTNFKSNS NQVALKNGSS SQHLFNASAN VEARYYYGDT SYFYMNAGVL QEFANFGSSN AVSLNTFKVN AAHNPLSTHA RVMMGGELKL AKEVFLNLGF VYLHNLISNI GHFASNLGMR YSF | |
Greater than 90% as determined by SDS-PAGE. |
899415 | |
10 μg | |
Not determined. | |
Full Length | |
H. pylori | |
His-B2M | |
Tris-based buffer, 50% glycerol |