missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Feline SDF 1b Protein
A cDNA sequence encoding the SDF 1b was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10862-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
493806 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity as determined by its ability to chemoattract human peripheral T cells stimulated with IL-2 and PHA in a concentration range from 10-75 ng/ml corresponding to a Specific Activity of 13,334-100,000IU/mg. | |
Feline | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives. |
SDF 1b Protein | |
Research Use Only | |
CXCL12 | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRF KM | |
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |