missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Equine IL 2 Protein
A cDNA sequence encoding the IL 2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10700-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
100034204 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cell proliferation assay using murine CTLL-2 cells is less than 1.0 microg/ml, corresponding to a specific activity of > 1000 IU/mg. | |
Equine | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 6.0. |
IL 2 Protein | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
E. coli | |
APTSSSKRET QQQLKQLQMD LKLLLEGVNN NKNPKLSKML TFKINMPKKA TELKHLQCLE EELKPLEEML KNFLSKDIKE LMSNINVTVL GLKGSETRFT CEYDDETGTI VEFLNKWITF SQSIFSTMT | |
Greater than 97.0% as determined by SDS-PAGE and HPLC analyses. |