missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli TRXR Protein

A cDNA sequence encoding the TRXR was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10894-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15995488

  • 3120.00€ / 1 miligramo
Fecha estimada de envío: 25-07-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Especificaciones

Especificaciones

TRXR Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGTTKHSKLL ILGSGPAGYT AAVYAARANL QPVLITGMEK GGQLTTTTEV ENWPGDPNDL TGPLLMERMH EHATKFETEI IFDHINKVDL QNRPFRLNGD NGEYTCDALI IATGASARYL GLPSEEAFKG RGVSACATCD GFFYRNQKVA VIGGGNTAVE EALYLSNIAS EVHLIHRRDG FRAEKILIKR LMDKVENGNI ILHTNRTLEE VTGDQMGVTG VRLRDTQNSD NIESLDVAGL FVAIGHSPNT AIFEGQLELE NGYIKVQSGI HGNATQTSIP GVFAAGDVMD HIYRQAITSA GTGCMAALDA ERYLDGLADA K
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Specific activity is 4-5 units/ml, and was measured in a coupled assay with DTNB and NADPH. The amount of TNB generated by NADPH was measured in absorbance at 412 nm.
E. coli
Untagged
TRXR E. Coli solution containing 20mM Tris HCl pH 8.0, 1mM DTT, and 10% glycerol.
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales