missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli LexA repressor Protein
A DNA sequence encoding the Escherichia coli (strain K12) LexA repressor, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7390-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_418467.1 | |
LexA repressor Protein | |
Research Use Only | |
lexA | |
Recombinant Protein | |
E. coli | |
MKALTARQQE VFDLIRDHIS QTGMPPTRAE IAQRLGFRSP NAAEEHLKAL ARKGVIEIVS GASRGIRLLQ EEEEGLPLVG RVAAGEPLLA QQHIEGHYQV DPSLFKPNAD FLLRVSGMSM KDIGIMDGDL LAVHKTQDVR NGQVVVARID DEVTVKRLKK QGNKVELLPE NSEFKPIVVD LRQQSFTIEG LAVGVIRNGD WL | |
Greater than 90% as determined by SDS-PAGE. |
948544 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |