missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli G6PD Protein

A cDNA sequence encoding the G6PD was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10622-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15994438

Cantidad Precio
1 194.62€ / 50 microgramos
10 182.86€ / 50 microgramos
Envío estimado: 10-03-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.



Descripción

Descripción

Especificaciones

Especificaciones

2539
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
G6PD
Recombinant Protein
E. coli
MAVTQTAQAC DLVIFGAKGD LARRKLLPSL YQLEKAGQLN PDTRIIGVGR ADWDKAAYTK VVREALETFM KETIDEGLWD TLSARLDFCN LDVNDTAAFS RLGAMLDQKN RITINYFAMP PSTFGAICKG LGEAKLNAKP ARVVMEKPLG TSLATSQEIN DQVGEYFEEC QVYRIDHYLG KETVLNLLAL RFANSLFVNN WDNRTIDHVE ITVAEEVGIE GRWGYFDKAG QMRDMIQNHL LQILCMIAMS PPSDLSADSI RDEKVKVLKS LRRIDRSNVR EKTVRGQYTA GFAQGKKVPG YLEEEGANKS SNTETFVAIR VDIDNWRWAG VPFYLRTGKR LPTKCSEVVV YFKTPELNLF KESWQDLPQN KLTIRLQPDE GVDIQVLNKV PGLDHKHNLQ ITKLDLSYSE TFNQTHLADA YERLLLETMR GIQALFVRRD EVEEAWKWVDSITEAWAMDN DAPKPYQAGT WGPVASVAMI TRDGRSWNEF E
G6PD Protein
50 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity > 70 units/mg obtained by measuring the increase of beta-NADPH in absorbance at 340 nm resulting from the reduction beta- NADP. One unit oxidizes 1.0 umole D-glucose-6-phosphate to 6-phospho-D-gluconate per min in the presence of beta-NADP at pH 7.4 at 25C.
E. coli
Untagged
The G6PD protein contains 50mM MES 6.0, 0.1mM PMSF, 2mM EDTA, 0.5mM DTT and 10% glycerol.
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales