missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Cathepsin K Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Cathepsin K, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5888-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001029168.1 | |
Cathepsin K Protein | |
Research Use Only | |
Ctsk | |
Recombinant Protein | |
E. coli | |
APDSVDYRKK GYVTPVKNQG QCGSCWAFSS VGALEGQLKK KTGKLLNLSP QNLVDCVSEN DGCGGGYMTN AFQYVQKNRG IDSEDAYPYV GQDESCMYNP TGKAAKCRGY REIPEGNEKA LKRAVARVGP ISVAIDASLT SFQFYSKGVY YDENCNSDNL NHAVLAVGYG IQKGNKHWII KNSWGENWGN KGYILMARNK NNACGIANLA SFPKM | |
Greater than 90% as determined by SDS-PAGE. |
608843 | |
200 μg | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |