Learn More
enQuireBio™ Recombinant Dengue virus Serotype 2 Envelope Protein
Recombinant Protein
Marca: enQuireBio™ QP10477-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Serotype 2 Envelope Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
PBS pH7.4. |
500 μg | |
Untested | |
Dengue virus (Strain: Thailand/16681/1984) | |
His | |
>90% as determined by SDS-PAGE. |