Learn More
enQuireBio™ Recombinant Dengue virus Dengue virus serotype 4 Envelope Protein
Recombinant Protein
Marca: enQuireBio™ QP10482-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
serotype 4 Envelope Protein | |
500 μg | |
Untested | |
Dengue virus | |
His |
>90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Human | |
PBS pH7.4. |