missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Vitellogenin-2 Protein
A DNA sequence encoding the Gallus gallus (Chicken) Vitellogenin-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8859-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001026447.1 | |
Vitellogenin-2 Protein | |
Research Use Only | |
VTG2 | |
Recombinant Protein | |
E. coli | |
SRRSYLYNYE GSMLNGLQDR SLGKAGVRLS SKLEISGLPE NAYLLKVRSP QVEEYNGVWP RDPFTRSSKI TQVISSCFTR LFKFEYSSGR IGNIYAPEDC PDLCVNIVRG ILNMFQMTIK KSQNVYELQE AGIGGICHAR YVIQEDRKNS RIYVTRTVDL NNCQEKVQKS IGMAYIYPCP VDVMKERLTK GTTAFSYKLK QSDSGTLITD VSSRQVYQIS PFNEPTGVAV MEARQQLTLV EVRSERGSAP DVPMQNYGSL RYRFPAVLPQ MPLQLIKTKN PEQRIVETLQ HIVLNNQQDF HDDVSYRFLE VVQLCRIANA DNLESIWRQV SDKPRYRRWL LSAVSASGTT ETLKFLKNRI RNDDLNYIQT LLTVSLTLHL LQADEHTLPI AADLMTSSRI QKNPVLQQVA CLGYSSVVNR YCSQTSACPK EALQPIHDLA DEAISRGRED KMKLALKCIG NMGEPASLKR ILKFLPISSS SAADIPVHIQ IDAITALKKI AWKDPKTVQG YLIQILADQS LPPEVRMMAC AVIFETRPAL ALITTIANVA MKESNMQVAS FVYSHMKSLS KSRLPFMYNI SSACNIALKL LSPKLDSMSY RYSKVIRADT YFDNYRVGAT GEIFVVNS | |
Greater than 90% as determined by SDS-PAGE. |
424533 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |