Learn More
enQuireBio™ Recombinant Bovine Adiponectin / Acrp30 / ADIPOQ Protein
A DNA sequence encoding the Bos taurus (Bovine) Adiponectin / Acrp30 / ADIPOQ, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7647-ye-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_777167.1 | |
Adiponectin / Acrp30 / ADIPOQ Protein | |
100 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
282865 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ADIPOQ | |
Recombinant Protein | |
Yeast | |
EDNMEDPPLP KGACAGWMAG IPGHPGHNGT PGRDGRDGTP GEKGEKGDPG LVGPKGDTGE TGITGIEGPR GFPGTPGRKG EPGESAYVYR SAFSVGLERQ VTVPNVPIRF TKIFYNQQNH YDGTTGKFLC NIPGLYYFSY HITVYLKDVK VSLYKNDKAL LFTHDQFQDK NVDQASGSVL LYLEKGDQVW LQVYEGENHN GVYADNVNDS TFTGFLLYHN IVE |