missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rac1 Antibody (CPTC-RAC1-1), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-08939AF647
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Rac1 Monoclonal antibody specifically detects Rac1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Especificaciones
Rac1 | |
Monoclonal | |
Alexa Fluor 647 | |
Cell migration-inducing gene 5 protein, MGC111543, p21-Rac1migration-inducing gene 5, Ras-like protein TC25, ras-related C3 botulinum toxin substrate 1, ras-related C3 botulinum toxin substrate 1 (rho family, small GTP bindingprotein Rac1), rho family, small GTP binding protein Rac1, TC25, TC-25 | |
Recombinant human full-length Rac1protein (Uniprot: P63000) | |
100 μg | |
Breast Cancer, Cancer, mTOR Pathway | |
5879 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CPTC-RAC1-1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Rac1 Antibody (CPTC-RAC1-1), Alexa Fluor™ 647, Novus Biologicals™ > 100 μg; Alexa Fluor 647
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido