Learn More
ACROBiosystems Rabbit BCMA / TNFRSF17 Protein, His Tag
Marca:  ACROBiosystems BCA-R52H4-100ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17206070
  • 419.88€ / 100 microgramos
Fecha estimada de envío: 11-04-2024
para ver el stock
Descripción
Descripción
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.5 kDa.
Especificaciones
Especificaciones
7.5 kDa
Rabbit BCMA, His Tag (BCA-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 1 - Asn 52 (Accession # G1SFN4-1).
-20°C
BCMA
Rabbit
HEK293
Bio-Markers & CD Antigens
100 μg
Gln 1 - Asn 52
TNFRSF17,CD269,BCM,BCMA
Unconjugated
His Tag
Powder