missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TICAM2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of TICAM2.
Marca: Abnova PAB19153.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TIRP is a Toll/interleukin-1 receptor (IL1R; MIM 147810) (TIR) domain-containing adaptor protein involved in Toll receptor signaling (see TLR4; MIM 603030).[supplied by OMIM
Especificaciones
TICAM2 | |
Polyclonal | |
Unconjugated | |
Western blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
TICAM2 | |
TICAM2 | |
A synthetic peptide corresponding near C-terminus of human TICAM2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody rasied against synthetic peptide of TICAM2. | |
In PBS (0.02% sodium azide) | |
MGC129876/MGC129877/TICAM-2/TIRAP3/TIRP/TRAM | |
Rabbit | |
Ion exchange chromatography purification | |
RUO | |
353376 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
TICAM2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™ > 100μg; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido