Learn More
SYT6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant SYT6.
Marca: Abnova PAB1224.100uL
Descripción
Synaptotagmins, such as SYT6, share a common domain structure that includes a transmembrane domain and a cytoplasmic region composed of 2 C2 domains. Some synaptotagmins are involved in synaptic membrane fusion, while others have a more general function in endocytosis. For further information on synaptotagmins, see MIM 185605.[supplied by OMIM
Especificaciones
SYT6 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant SYT6. | |
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide) | |
SYT6 | |
Recombinant His fusion protein corresponding to amino acids 87-510 of human SYT6. | |
RUO | |
148281 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1:1800) The optimal working dilution should be determined by the end user. | |
SYT6 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only