missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUSC2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RUSC2.
Marca: Abnova PAB25783.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
RUSC2 | |
Polyclonal | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
RUSC2 | |
RUSC2 | |
A synthetic peptide corresponding to 18 amino acids at N-terminus of human RUSC2. | |
100 μg | |
Primary | |
At least three isoforms are known to exist; this antibody will detect all three isoforms. This antibody is predicted to not cross-react with RUSC1. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of RUSC2. | |
In PBS (0.02% sodium azide) | |
Iporin/KIAA0375 | |
Rabbit | |
Peptide affinity purification | |
RUO | |
9853 | |
Human, Mouse | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
RUSC2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™ > 100μg; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido