missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU5F1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of POU5F1.
Marca: Abnova PAB12773.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
POU5F1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of POU5F1. | |
In TBS, pH 7.2 (BSA, 10% Proclin300) | |
POU5F1 | |
A synthetic peptide corresponding to internal domain of human, mouse and rat POU5F1. | |
RUO | |
5460/18999/294562 | |
Human, Mouse, Rat | |
Liquid |
Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
MGC22487/OCT3/OCT4/OTF3/OTF4 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes ∼39 KDa of POU5F1. It does not cross react with POU2F1, POU2F2. | |
Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
POU5F1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™ > 100μg; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido