missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Pig IgG whole molecule, FITC, Polyclonal, Secondary Antibody, Abnova™
Marca: Abnova PAB10793.2mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Rabbit anti-Pig IgG whole molecule | |
Polyclonal | |
Rabbit anti-pig IgG recognizes pig IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein. | |
Lyophilized from 5mM sodium phosphate, 70mM NaCl, 50% (v/v) glycerol, pH 7.4 | |
Pig IgG whole molecule | |
2 mg | |
Pig | |
Lyophilized |
Flow Cytometry, Immunofluorescence | |
FITC | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Rabbit anti-Pig IgG whole molecule, FITC, Polyclonal, Secondary Antibody, Abnova™ > 2mg; FITC
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido