missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Pig IgG F(ab')2 fragment,Polyclonal Secondary Antibody, Abnova™
Marca: Abnova PAB9474.2mL
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Rabbit anti-Pig IgG F(ab')2 fragment | |
Polyclonal | |
Rabbit anti-pig IgG recognizes pig IgG F(ab')2 fragment. This secondary antibody was purified using delipidation and defibrination. | |
Lyophilized from 10mM sodium phosphate, 250mM NaCl, pH 7.4 (0.01% (w/v) sodium azide) | |
Pig IgG F(ab')2 fragment | |
2 mL | |
Pig | |
Lyophilized |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against pig IgG F(ab')2 and pig serum. | |
Secondary | |
Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Rabbit anti-Pig IgG F(ab')2 fragment,Polyclonal Secondary Antibody, Abnova™ > 2mL; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido