missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Pig IgG F(ab')2 fragment, Biotin, Polyclonal, Secondary Antibody, Abnova™
Marca: Abnova PAB10801.2mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Rabbit anti-Pig IgG F(ab')2 fragment | |
Polyclonal | |
Rabbit anti-pig IgG recognizes pig IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin. | |
Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 | |
Pig IgG F(ab')2 fragment | |
2 mg | |
Pig | |
Lyophilized |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Biotin | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Rabbit anti-Pig IgG F(ab')2 fragment, Biotin, Polyclonal, Secondary Antibody, Abnova™ > 2mg; Biotin
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido