missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Pig IgG F(ab')2 fragment, AP, Polyclonal, Secondary Antibody, Abnova™
Marca: Abnova PAB10800.1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Rabbit anti-Pig IgG F(ab')2 fragment | |
Polyclonal | |
Rabbit anti-pig IgG recognizes pig IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Alkaline Phosphatase. | |
In 50mM Tris-Cl, 150mM NaCl, 1mM MgCl, 0.1mM ZnCl2, 50% (v/v) glycerol, pH 8.0 (0.01% sodium azide) | |
Pig IgG F(ab')2 fragment | |
1 mg | |
Pig | |
Liquid |
Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
Alkaline Phosphatase (AP) | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Store at 4°C. Do not Freeze. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Rabbit anti-Pig IgG F(ab')2 fragment, AP, Polyclonal, Secondary Antibody, Abnova™ > 1mg; AP
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido