Learn More
MKI67 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MKI67.
Marca: Abnova PAB12698.100ug
Descripción
This gene encodes a nuclear protein that is associated with and may be necessary for cellular proliferation. Alternatively spliced transcript variants have been described. A related pseudogene exists on chromosome X. [provided by RefSeq
Especificaciones
MKI67 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MKI67. | |
In TBS, pH 7.2 (BSA, 10% Proclin300) | |
KIA/Ki-67 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes ∼67 KDa of MKI67. | |
Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. Upon initial thawing apportion into working aliquots and store at -80°C for up to six months. |
ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
MKI67 | |
MKI67 | |
A synthetic peptide corresponding to human MKI67. | |
RUO | |
4288 | |
Human, Mouse, Rat | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only