Learn More
IRS2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IRS2.
Brand: Abnova PAB12697.100ug
Description
This gene encodes the insulin receptor substrate 2, a cytoplasmic signaling molecule that mediates effects of insulin, insulin-like growth factor 1, and other cytokines by acting as a molecular adaptor between diverse receptor tyrosine kinases and downstream effectors. The product of this gene is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation, as well as by an interleukin 4 receptor-associated kinase in response to IL4 treatment. [provided by RefSeq
Specifications
IRS2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of IRS2. | |
In TBS, pH 7.2 (BSA, 10% Proclin300) | |
IRS2 | |
A synthetic peptide corresponding to residues surrounding S731 of human IRS2. | |
RUO | |
8660 | |
Human, Mouse, Rat | |
Liquid |
Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
IRS2 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes ∼180 KDa of human IRS2. | |
Store at 4°C. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only