Learn More
Rabbit anti-Horse IgG F(c) fragment, FITC, Polyclonal, Secondary Antibody, Abnova™
Marca: Abnova PAB10673.1500ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Rabbit anti-Horse IgG F(c) fragment | |
Polyclonal | |
Rabbit anti-horse IgG recognizes horse IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein. | |
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
Horse IgG F(c) fragment | |
1500 μg | |
Equine | |
Lyophilized |
Flow Cytometry, Immunofluorescence | |
FITC | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Título del producto
Rabbit anti-Horse IgG F(c) fragment, FITC, Polyclonal, Secondary Antibody, Abnova™ > 1500μg; FITC
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido