Learn More
FAM3C, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant FAM3C.
Marca: Abnova PAB0316.100ug
Descripción
This gene is a member of the family with sequence similarity 3 (FAM3) family and encodes a secreted protein with a GG domain. A change in expression of this protein has been noted in pancreatic cancer-derived cells. Alternate transcriptional splice variants which encode the same protein have been characterized. [provided by RefSeq
Especificaciones
FAM3C | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant FAM3C. | |
In PBS (0.2% gelatin, 0.05% sodium azide) | |
NP_055703 | |
FAM3C | |
Recombinant protein corresponding to amino acids 40-80 of human FAM3C. | |
RUO | |
10447 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
FAM3C | |
GS3786/ILEI | |
Rabbit | |
100 μg | |
Primary | |
Bovine, Canine, Chicken, Human, Monkey | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only