missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epb41l2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Epb4.1l2.
Marca: Abnova PAB15751.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Epb41l2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant Epb4.1l2. | |
Lyophilized from 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.05% sodium azide) | |
Recombinant GST fusion protein corresponding to 179 amino acids (E694-S872) of rat Epb4.1l2. | |
RUO | |
This antibody detects rat and mouse EPB41L2 protein. | |
Store at 4°C on dry atmosphere. After reconstitution with 200μL distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:25) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Rat | |
Lyophilized |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Epb41l2, Rabbit, Polyclonal Antibody, Abnova™ > 100μg; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido