Learn More
EBAG9 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EBAG9.
Marca: Abnova PAB12726.100ug
Descripción
This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element (ERE) found in the 5'-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5' UTR, but encoding the same protein, have been identified for this gene. [provided by RefSeq]
Especificaciones
EBAG9 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of EBAG9. | |
In TBS, pH 7.2 (BSA, 10% Proclin300) | |
EB9/PDAF/RCAS1 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes ∼32 KDa EBAG9. | |
Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
EBAG9 | |
EBAG9 | |
A synthetic peptide corresponding to C-terminus of human EBAG9. | |
RUO | |
9166 | |
Human | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only