Learn More
DCLK1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of DCLK1.
Marca: Abnova PAB19206.100ug
Descripción
DCAMKL1 is a microtubule-associated protein that phosphorylates itself and myelin basic protein (MBP; MIM 159430). DCAMKL1 has microtubule polymerizing activity that is independent of its protein kinase activity (Lin et al., 2000 [PubMed 11124993]).[supplied by OMIM
Especificaciones
DCLK1 | |
Polyclonal | |
Unconjugated | |
Western blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
DCLK1 | |
DCAMKL1/DCDC3A/DCLK/KIAA0369 | |
Rabbit | |
Affinity purification | |
RUO | |
9201 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody rasied against synthetic peptide of DCLK1. | |
In PBS (0.02% sodium azide) | |
NP_004725 | |
DCLK1 | |
A synthetic peptide corresponding to 14 amino acids near N-terminus of human DCLK1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only