missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Col2a1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Col2a1.
Marca: Abnova PAB19147.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Col2a1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against native Col2a1. | |
In PBS | |
Col2/Col2a/Col2a-1/Del1/Dmm/MGC90638 | |
Rabbit | |
Affinity purification | |
RUO | |
12824 | |
Human, Mouse, Rat | |
Liquid |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western blot (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
Col2a1 | |
Col2a1 | |
Native purified Col2a1 CnBr fragment from mouse chondrosarcoma. | |
100 μg | |
Primary | |
It exhibits very moderately with bovine collagen type XI and slightly with collagen type I (< 10%). | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Col2a1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™ > 100μg; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido