missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPF, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant CENPF.
Marca: Abnova PAB12373.50uL
This item is not returnable.
View return policy
Specifications
CENPF | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant CENPF. | |
In PBS (50% glycerol, 2.0mg/mL BSA, 0.05% sodium azide) | |
Rabbit | |
50 μL | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Flow Cytometry (1:50-1:200) Immunocytochemistry (1:200-1:1000) Immunofluorescence (1:750) Immunoprecipitation (1:50) Western Blot (1:1500-1:2500) The optimal working dilution should be determined by the end user. | |
P49454 | |
Recombinant protein corresponding to C-terminus of CENPF. | |
RUO | |
This antibody is specific to CENP-F. | |
Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CENPF, Rabbit, Polyclonal Antibody, Abnova™ > 50μL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction