missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calr Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Calr.
Marca: Abnova PAB12147.100uL
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Calr | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant Calr. | |
In PBS (0.05% sodium azide) | |
P14211 | |
Calr | |
Recombinant fusion protein corresponding to mouse Calr. | |
RUO | |
12317 | |
Bovine, Human, Mouse, Rat | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Immunocytochemistry (1:250) Immunofluorescence (1:50) Immunohistochemistry (1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Calr | |
CRT/Calregulin | |
Rabbit | |
100 μL | |
Primary | |
This antibody is specific to calreticulin. | |
Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Calr Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal Antibody, Abnova™ > 100μL; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido