Learn More
BCL2L1 Rabbit anti-Dog, Gerbils, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a recombinant BCL2L1.
Marca: Abnova PAB0237.50uL
Descripción
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The proteins encoded by this gene are located at the outer mitochondrial membrane, and have been shown to regulate outer mitochondrial membrane channel (VDAC) opening. VDAC regulates mitochondrial membrane potential, and thus controls the production of reactive oxygen species and release of cytochrome C by mitochondria, both of which are the potent inducers of cell apoptosis. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported. The longer isoform acts as an apoptotic inhibitor and the shorter form acts as an apoptotic activator. [provided by RefSeq
Especificaciones
BCL2L1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a recombinant BCL2L1. | |
In serum (0.05% sodium azide) | |
BCL-XL/S/BCL2L/BCLX/Bcl-X/DKFZp781P2092/bcl-xL/bcl-xS | |
Rabbit | |
50 μL | |
Primary | |
Canine, Gerbil, Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
BCL2L1 | |
BCL2L1 | |
Recombinant protein corresponding to human BCL2L1 . | |
RUO | |
598 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only