Learn More
BAG3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant BAG3.
Marca: Abnova PAB0330.50uL
Descripción
BAG proteins compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. All the BAG proteins have an approximately 45-amino acid BAG domain near the C terminus but differ markedly in their N-terminal regions. The protein encoded by this gene contains a WW domain in the N-terminal region and a BAG domain in the C-terminal region. The BAG domains of BAG1, BAG2, and BAG3 interact specifically with the Hsc70 ATPase domain in vitro and in mammalian cells. All 3 proteins bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner. [provided by RefSeq
Especificaciones
BAG3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant BAG3. | |
In serum (0.05% sodium azide) | |
BAG-3/BIS/CAIR-1/MGC104307 | |
Rabbit | |
50 μL | |
Primary | |
Human, Mouse | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
BAG3 | |
BAG3 | |
Recombinant protein corresponding to C-terminus 196 amino acids of human BAG3. | |
RUO | |
9531 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Seguridad y manipulación
- BAG3 polyclonal antibody
Palabra de advertencia
- Atención
Clasificación
- Lesión ocular grave/irritación ocular Categoría 2
- Corrosión o irritación cutáneas Categoría 2
Indicaciones de peligro
- H315-Provoca irritación cutánea.
- H319-Provoca irritación ocular grave.
Consejos de prudencia
- P260-No respirar el polvo/el humo/el gas/la niebla/los vapores/el aerosol.
- P305+P351+P338-EN CASO DE CONTACTO CON LOS OJOS: Aclarar cuidadosamente con agua durante varios minutos. Quitar las lentes de contacto, si lleva y resulta fácil. Seguir aclarando.
- P310-Llamar inmediatamente a un CENTRO DE TOXICOLOG A/médico/.
Otros peligros
- MIXTURE LIST-Contém: sodium azide
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only