Learn More
ASAH1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ASAH1.
Marca: Abnova PAB13056.100ug
Descripción
This gene encodes a heterodimeric protein consisting of a nonglycosylated alpha subunit and a glycosylated beta subunit that is cleaved to the mature enzyme posttranslationally. The encoded protein catalyzes the synthesis and degradation of ceramide into sphingosine and fatty acid. Mutations in this gene have been associated with a lysosomal storage disorder known as Farber disease. Multiple transcript variants encoding several distinct isoforms have been identified for this gene. [provided by RefSeq
Especificaciones
ASAH1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ASAH1. | |
In PBS (0.02% sodium azide) | |
EAW63795 | |
ASAH1 | |
A synthetic peptide corresponding to C-terminus 16 amino acids of human ASAH1. | |
RUO | |
427 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
ASAH1 | |
AC/ASAH/FLJ21558/FLJ22079/PHP/PHP32 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only