Learn More
Invitrogen™ Rab4 Recombinant Rabbit Monoclonal Antibody (24GB825)
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA557661
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Rab4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Protein transport. Probably involved in vesicular traffic.
Especificaciones
Rab4 | |
Recombinant Monoclonal | |
Unconjugated | |
Rab4a | |
AI848268; HRES1; HRES-1; HRES-1/RAB4; HTLV-1 related endogenous sequence; RAB4; Rab4a; RAB4A, member RAS oncogene family; ras-related protein Rab-4A; Ras-related small GTP binding protein 4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
19341, 25532, 5867 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
24GB825 | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P05714, P20338, P56371 | |
Rab4a | |
A synthesized peptide derived from human Rab4 (aa 50-75). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Título del producto
Invitrogen™ Rab4 Recombinant Rabbit Monoclonal Antibody (24GB825) > 20 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido