Learn More
Tuttnauer R.PCR Remote PC Software
Remote PC Reporting, PC software for generating reports which include: graph of the cycle data, numeric data of the cycle, copy of cycle print-outs, table of all measured values, table of all parameters and option to save all reports in PDF file format.
Marca:  Tuttnauer SRV000-0569
Código de producto. 17809598
  • 615.00€ / cada uno
Solo quedan null
Fecha estimada de envoi: 03-07-2025
Descripción
  • PC software for generating reports which include: graph of the cycle data, numeric data of the cycle, copy of cycle print-outs, table of all measured values, table of all parameters and option to save all reports in PDF file format.
The software allows 2 modes of operation:
  • Online mode:
    • Up to 8 autoclaves may be accessed using a PC within the organization via an Ethernet connection.
    • These autoclaves can each be remotely monitored at the same time showing all reports and graphs.
    • History files are automatically downloaded from the autoclave.
  • Offline mode:
    • Data is transferred from the autoclave using a standard USB memory device.
    • This data can then be viewed with the report generator software on a PC.
Especificaciones
Especificaciones
Software
Documentos
One moment while we fetch your results.
Certificados
Promociones
Promociones
Título del producto
Tuttnauer R.PCR Remote PC Software > R.PCR Remote PC Software