Learn More
R&D Systems™ Recombinant Mouse Inhibin A Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca: R&D Systems™ 8346-IN-010/CF

Descripción
Measured by its ability to neutralize Activin-mediated erythroid differentiation of K562 human chronic myelogenous leukemia cells. The ED50 for this effect is 5-30 ng/mL.Especificaciones
CAA49324 (alpha chain) & P08476 (beta chain) | |
3623 | |
Inhibin A | |
SDS-PAGE: 12-14kDa & 17-20kDa, reducing conditions | |
Chinese Hamster Ovary cell line, CHO-derived mouse Inhibin A protein Ser234-Ile366 (alpha chain) & Gly311-Ser426 (beta chain) | |
RUO | |
Carrier Protein | |
<0.10 EU / 1 µg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 100 µg/mL in 4 mM HCl. |
Lyophilized from a 0.2 µm filtered solution in HCl. | |
M.W. (Observed): 12-14 kDa & 17-20 kDa, reducing conditions; M.W. (theoretical): 14.5 kDa (alpha chain) & 13.0 kDa (beta chain) | |
Reconstitute at 100μg/mL in 4mM HCl. | |
10 µg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Ser234 (α chain) & Gly311 (β chain) | |
Disulfide-linked heterodimer | |
Inha | |
Yes |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.