Learn More
R&D Systems™ Human XCL2/SCM-1 beta Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca: R&D Systems™ 8418-X2-025

Descripción
The Recombinant Human XCL2/SCM-1 beta Protein is derived from HEK293. The Recombinant Human XCL2/SCM-1 beta Protein has been validated for the following applications: Bioactivity.Especificaciones
Q9UBD3 | |
6846 | |
XCL2/SCM-1 beta | |
SDS-PAGE: 19-22kDa, reducing conditions | |
Human embryonic kidney cell,HEK293-derived human XCL2/SCM-1 beta protein Val22-Gly114 | |
RUO | |
Unconjugated | |
Reconstitute at 250 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein | |
10kDa | |
Reconstitute at 250μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Val22 | |
Yes |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.