Learn More
Pyrex™ Borosilicate Glass Square Media-lab Bottles
Manufactured from Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Square Media-lab Bottles allow closer stacking in water baths or on shelves. Square shape provides up to 20% saving of space compared to round bottles. Each bottle includes a polypropylene cap and pouring ring. For use up to 140° C.
Marca:  Pyrex 1520/08D
Código de producto. 10527860
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Descripción
Descripción
  • Square shape allows closer stacking in water baths or on shelves
  • Up to 20% saving of space compared to round bottles
  • Bottle complete with polypropylene cap and pouring ring
  • For use up to 140°C
  • Printed with retrace code: downloadable batch certificate
Certificados
Promociones
Promociones
Título del producto
Pyrex™ Borosilicate Glass Square Media-lab Bottles > Capacity: 500mL; Dimensions: 78 dia. x 181mmH; Each