Learn More
DiscoverX PrecisION™ hnAChR α3/β4 Recombinant Stable Cell Line
The PrecisION™ hnAChR α3/β4 Recombinant Stable Cell Line measures nAChR α3/β4 ion channel activity via electrophysiological measurements. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX CYL3057
Descripción
The PrecisION™ hnAChR α3/β4 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying nAChR α3/β4 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses nAChR α3/β4 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells.Especificaciones
Ion Channel Activity | |
Cell Lines | |
nAChR α3/β4 |
293H | |
2 viales | |
M37981 |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.