Learn More
Phospho-ZIPK (Ser311) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 51021-1-AP-100UL
 Ver más versiones de este producto
Código de producto. 16876285
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Descripción
Descripción
This antibody is specific to ZIPK-Phospho 311Ser.
The protein encoded by this gene is a component of desmosomes and of the epidermal cornified envelope in keratinocytes. The N-terminal domain of this protein interacts with the plasma membrane and its C-terminus interacts with intermediate filaments. Through its rod domain, this protein forms complexes with envoplakin. This protein may serve as a link between the cornified envelope and desmosomes as well as intermediate filaments. AKT1/PKB, a protein kinase mediating a variety of cell growth and survival signaling processes, is reported to interact with this protein, suggesting a possible role for this protein as a localization signal in AKT1-mediated signaling.
Especificaciones
Especificaciones
Phospho-ZIPK (Ser311)
Polyclonal
Unconjugated
DAPK3
DAP kinase 3, DAP like kinase, DAPK3, Dlk, ZIP, ZIP kinase, ZIPK
Rabbit
Antigen Affinity Chromatography
RUO
13144, 1613
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43293, O54784
Dapk3
Peptide
100 μL
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
One moment while we fetch your results.
Certificados
Promociones
Promociones
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Phospho-ZIPK (Ser311) Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 100 μL; Unconjugated