missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-alpha Synuclein (Tyr136) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA537743
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
A suggested positive control for Western blot is mouse brain cells; suggested positive control for ICC/IF is Hela cells.
Phospho-alpha Synuclein (Tyr136) Polyclonal antibody specifically detects Phospho-alpha Synuclein (Tyr136) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Especificaciones
Phospho-alpha Synuclein (Tyr136) | |
Polyclonal | |
Unconjugated | |
SNCA | |
Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor) | |
Rabbit | |
Affinity chromatography | |
RUO | |
20617, 29219, 6622 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O55042, P37377, P37840 | |
SNCA | |
Peptide sequence around phosphorylation site of tyrosine 136 (Q-D-Y(p)-E-P) derived from Human alpha-Synuclein. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Phospho-alpha Synuclein (Tyr136) Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido