Learn More
PD-L2 (PDCD1LG2) Mouse anti-Human, Clone: OTI10G2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF806575

Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD273 (PDCD1 Ligand 2, Programmed cell death 2 ligand 2, PD-L2, B7-DC) is involved in cell-mediated immune responses initiated by T lymphocytes that are stimulated by cognate peptides bound to MHC molecules on antigen-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as CD273 that mediate inhibitory signals from the APC (antigen presenting cell). CD273 can bind two different but related ligands, PDL-1 and PDL-2, both of which are thought act as a negative regulator of T cell activation. However, it has been suggested that PDL-2 can act to stimulate an immunogenic response through and alternative receptor from PD-1. At least three isoforms of PDL-2 are known to exist; CD273 is specific to the largest isoform. Diseases associated with CD273 include cysticercosis, and myasthenic syndrome.Especificaciones
PD-L2 (PDCD1LG2) | |
Monoclonal | |
Unconjugated | |
PDCD1LG2 | |
B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
80380 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI10G2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BQ51 | |
PDCD1LG2 | |
Full length human recombit protein of human PDCD1LG2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.